missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromo-4,5-dichloroimidazole, 98%

Manufacturer:  Alfa Aesar™ A1489614

 View more versions of this product

Catalog No. AAA1489614



Specifications

Specifications

2-Bromo-4,5-dichloroimidazole
~215°C (decomposition)
16076-27-0
MFCD00067894
2-bromo-4,5-dichloroimidazole, imidazole, 2-bromo-4,5-dichloro, 1h-imidazole, 2-bromo-4,5-dichloro, imidazole,5-dichloro, acmc-20ao2s, 1h-imidazole,5-dichloro, 5-23-04-00463 beilstein handbook reference, 2-bromo-4,5-dichloro-imidazole, 2-bromo-4,5-dichloro-3h-imidazole, 2-bromo-4,5-dichloroimidazole
C1(=C(N=C(N1)Br)Cl)Cl
215.859
215.87
Odorless
25g
C3HBrCl2N2
606732
SULQOTGFPPFUMG-UHFFFAOYSA-N
2-bromo-4,5-dichloro-1H-imidazole
99484
98%

Chemical Identifiers

16076-27-0
215.859
SULQOTGFPPFUMG-UHFFFAOYSA-N
99484
C1(=C(N=C(N1)Br)Cl)Cl
C3HBrCl2N2
MFCD00067894
2-bromo-4,5-dichloroimidazole, imidazole, 2-bromo-4,5-dichloro, 1h-imidazole, 2-bromo-4,5-dichloro, imidazole,5-dichloro, acmc-20ao2s, 1h-imidazole,5-dichloro, 5-23-04-00463 beilstein handbook reference, 2-bromo-4,5-dichloro-imidazole, 2-bromo-4,5-dichloro-3h-imidazole, 2-bromo-4,5-dichloroimidazole
2-bromo-4,5-dichloro-1H-imidazole
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning

RTECSNumber : NI3517500

SDS Documents