Please login to your online account to display your discounted pricing

2-Bromo-4,5-dichloro-1H-imidazole, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 2-Bromo-4, 5-dichloro-1H-imidazole
CAS 16076-27-0
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C3HBrCl2N2
Formula Weight 215.86
MDL Number MFCD00067894
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

KM01467DA

  SDS
fisher scientific company
KM01467DA
Amber Glass Bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 215.86
Quantity 1g
Assay 97%
CAS Min % 97.0
Health Hazard 1 GHS Signal Word: Warning
CAS 16076-27-0
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Harmful in contact with skin.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if inhaled.
Assay Percent Range 97%
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD00067894
Packaging Amber Glass Bottle
Chemical Name or Material 2-Bromo-4, 5-dichloro-1H-imidazole
Molecular Formula C3HBrCl2N2