missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Bromo-3-(trifluoromethyl)pyridine, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H2582206

 View more versions of this product

Catalog No. AAH2582206



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Bromo-3-(trifluoromethyl)pyridine
37°C to 39°C
29°C (84°F)
MFCD00153084
2-bromo-3-trifluoromethyl pyridine, 2-bromo-3-trifluorormethyl pyridine, pyridine, 2-bromo-3-trifluoromethyl, 2-bromo-3-trifluormethyl-pyridine, pubchem2999, acmc-1bx8n, ksc174i2d, tpc-py049, buttpark 27\06-85
C1=CC(=C(N=C1)Br)C(F)(F)F
225.996
226
175205-82-0
76°C (8 mmHg)
C6H3BrF3N
5 g
OFGSIPQYQUVVPL-UHFFFAOYSA-N
2-bromo-3-(trifluoromethyl)pyridine
5193820
98%

Chemical Identifiers

175205-82-0
225.996
OFGSIPQYQUVVPL-UHFFFAOYSA-N
5193820
C1=CC(=C(N=C1)Br)C(F)(F)F
C6H3BrF3N
MFCD00153084
2-bromo-3-trifluoromethyl pyridine, 2-bromo-3-trifluorormethyl pyridine, pyridine, 2-bromo-3-trifluoromethyl, 2-bromo-3-trifluormethyl-pyridine, pubchem2999, acmc-1bx8n, ksc174i2d, tpc-py049, buttpark 27\06-85
2-bromo-3-(trifluoromethyl)pyridine
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P240-P241-P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P370+P378q-P501c

H228-H300-H315-H318-H335

DOTInformation : Hazard Class: 4.1; Packaging Group: III

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only