missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromo-3-(trifluoromethyl)benzoic acid, 98%

$31.04 - $362.80

Chemical Identifiers

CAS 177420-63-2
Molecular Formula C8H4BrF3O2
Molecular Weight (g/mol) 269.017
MDL Number MFCD08532493
InChI Key FIMDBAHLQCCYJK-UHFFFAOYSA-N
Synonym 2-bromo-3-carboxybenzotrifluoride, 2-bromo-3-trifluoromethyl benzoic acid, 2-bromo-3-trifluoromethylbenzoicacid, benzoic acid, 2-bromo-3-trifluoromethyl
PubChem CID 15261012
IUPAC Name 2-bromo-3-(trifluoromethyl)benzoic acid
SMILES C1=CC(=C(C(=C1)C(F)(F)F)Br)C(=O)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH63761MD
View Documents
Alfa Aesar™
H63761MD
250mg Each for $31.04
 
AAH6376103
View Documents
Alfa Aesar™
H6376103
1g Each for $91.45
 
AAH6376106
View Documents
Alfa Aesar™
H6376106
5g Each for $362.80
 
Description

Description

Applications
2-Bromo-3-(trifluoromethyl)benzoic acid is used as a intermediate for pharmaceutical and chemical research.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications

Chemical Identifiers

177420-63-2
269.017
FIMDBAHLQCCYJK-UHFFFAOYSA-N
15261012
C1=CC(=C(C(=C1)C(F)(F)F)Br)C(=O)O
C8H4BrF3O2
MFCD08532493
2-bromo-3-carboxybenzotrifluoride, 2-bromo-3-trifluoromethyl benzoic acid, 2-bromo-3-trifluoromethylbenzoicacid, benzoic acid, 2-bromo-3-trifluoromethyl
2-bromo-3-(trifluoromethyl)benzoic acid

Specifications

250mg
C8H4BrF3O2
2-bromo-3-carboxybenzotrifluoride, 2-bromo-3-trifluoromethyl benzoic acid, 2-bromo-3-trifluoromethylbenzoicacid, benzoic acid, 2-bromo-3-trifluoromethyl
FIMDBAHLQCCYJK-UHFFFAOYSA-N
2-bromo-3-(trifluoromethyl)benzoic acid
15261012
Solid
2-Bromo-3-(trifluoromethyl)benzoic acid
177420-63-2
MFCD08532493
Slightly soluble in water.
C1=CC(=C(C(=C1)C(F)(F)F)Br)C(=O)O
269.017
269.02
98%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures