missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromo-3-(trifluoromethyl)benzoic acid, 98%

Manufacturer:  Alfa Aesar™ H6376103

 View more versions of this product

Catalog No. AAH6376103



Description

Description

Applications
2-Bromo-3-(trifluoromethyl)benzoic acid is used as a intermediate for pharmaceutical and chemical research.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications

Specifications

2-Bromo-3-(trifluoromethyl)benzoic acid
177420-63-2
MFCD08532493
Slightly soluble in water.
C1=CC(=C(C(=C1)C(F)(F)F)Br)C(=O)O
269.017
269.02
98%
1g
C8H4BrF3O2
2-bromo-3-carboxybenzotrifluoride, 2-bromo-3-trifluoromethyl benzoic acid, 2-bromo-3-trifluoromethylbenzoicacid, benzoic acid, 2-bromo-3-trifluoromethyl
FIMDBAHLQCCYJK-UHFFFAOYSA-N
2-bromo-3-(trifluoromethyl)benzoic acid
15261012
Solid

Chemical Identifiers

177420-63-2
269.017
FIMDBAHLQCCYJK-UHFFFAOYSA-N
15261012
C1=CC(=C(C(=C1)C(F)(F)F)Br)C(=O)O
C8H4BrF3O2
MFCD08532493
2-bromo-3-carboxybenzotrifluoride, 2-bromo-3-trifluoromethyl benzoic acid, 2-bromo-3-trifluoromethylbenzoicacid, benzoic acid, 2-bromo-3-trifluoromethyl
2-bromo-3-(trifluoromethyl)benzoic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents