Please login to your online account to display your discounted pricing

(2-Bromo-3-thienyl)methylamine, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material (2-Bromo-3-thienyl)methylamine hydrochloride
CAS 157664-47-6
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C5H6BrNS·ClH
Formula Weight 228.54
MDL Number MFCD04971979
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

SPB08448CB

 
fisher scientific company
SPB08448CB
Amber Glass Bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 228.54
Quantity 250mg
Assay 97%
CAS Min % 97.0
Health Hazard 2 GHS H Statement:
Harmful in contact with skin.
Causes serious eye damage.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful if inhaled.
Chemical Name or Material (2-Bromo-3-thienyl)methylamine hydrochloride
Packaging Amber Glass Bottle
Assay Percent Range 97%
Molecular Formula C5H6BrNS·ClH
MDL Number MFCD04971979
CAS 157664-47-6
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.