Please login to your online account to display your discounted pricing

(2-Bromo-3-thienyl)methylamine, 97%, Maybridge


Quantity 250mg
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  SPB08448CB

 View more versions of this product

Catalog No. SPB08448CB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 228.54
Quantity 250mg
Assay 97%
CAS Min % 97.0
Health Hazard 2 GHS H Statement:
Harmful in contact with skin.
Causes serious eye damage.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful if inhaled.
Chemical Name or Material (2-Bromo-3-thienyl)methylamine hydrochloride
Packaging Amber Glass Bottle
Assay Percent Range 97%
Molecular Formula C5H6BrNS·ClH
MDL Number MFCD04971979
CAS 157664-47-6
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material (2-Bromo-3-thienyl)methylamine hydrochloride
CAS 157664-47-6
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C5H6BrNS·ClH
Formula Weight 228.54
MDL Number MFCD04971979
Accessories

GHS Signal Word: Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful in contact with skin.
Causes serious eye damage.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful if inhaled.