missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Bromo-3-pyridinol 99%, ACROS Organics™

Manufacturer:  Acros Organics 152170050

 View more versions of this product

Catalog No. AC152170050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

2-Bromo-3-pyridinol
Glass bottle
5g
6602-32-0
100.0
C5H4BrNO
MFCD00006220
YKHQFTANTNMYPP-UHFFFAOYSA-N
2-bromopyridin-3-ol
23070
Crystalline Powder
Authentic
Brown to Gray
99%
98.5
99%
Br(C5H3N)OH
2-bromo-3-hydroxypyridine, 2-bromo-3-pyridinol, 3-pyridinol, 2-bromo, 2-bromo-pyridin-3-ol, 3-hydroxy-2-bromopyridine, unii-9dab85h359, 2-bromo-3-hydroxy pyridine, 2-bromo-3-hydroxy-pyridine, 2-bromo-3-hydroxypyridine 2-bromo-3-pyridinol, 2-bromo3-pyridinol
C1=CC(=C(N=C1)Br)O
173.997
174
≥98.5%

Chemical Identifiers

6602-32-0
173.997
YKHQFTANTNMYPP-UHFFFAOYSA-N
23070
C1=CC(=C(N=C1)Br)O
C5H4BrNO
MFCD00006220
2-bromo-3-hydroxypyridine, 2-bromo-3-pyridinol, 3-pyridinol, 2-bromo, 2-bromo-pyridin-3-ol, 3-hydroxy-2-bromopyridine, unii-9dab85h359, 2-bromo-3-hydroxy pyridine, 2-bromo-3-hydroxy-pyridine, 2-bromo-3-hydroxypyridine 2-bromo-3-pyridinol, 2-bromo3-pyridinol
2-bromopyridin-3-ol
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Harmful in contact with skin.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 229-547-5

RTECSNumber : UU7705000

TSCA : TSCA

SDS Documents