missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Bromo-3-pyridinol, 99%, Thermo Scientific™

$327.06 - $327.06

Chemical Identifiers

CAS 6602-32-0
Molecular Formula C5H4BrNO
Molecular Weight (g/mol) 174.00
MDL Number MFCD00006220
InChI Key YKHQFTANTNMYPP-UHFFFAOYSA-N
Synonym 2-bromo-3-hydroxy pyridine, 2-bromo-3-hydroxy-pyridine, 2-bromo-3-hydroxypyridine, 2-bromo-3-hydroxypyridine 2-bromo-3-pyridinol, 2-bromo-3-pyridinol, 2-bromo-pyridin-3-ol, 2-bromo3-pyridinol, 3-hydroxy-2-bromopyridine, 3-pyridinol, 2-bromo, unii-9dab85h359
PubChem CID 23070
IUPAC Name 2-bromopyridin-3-ol
SMILES OC1=CC=CN=C1Br
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC152170250
SDS View Documents Promotion Details
Thermo Fisher Scientific
152170250
25g Glass bottle
Each for $327.06
 
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

6602-32-0
174.00
YKHQFTANTNMYPP-UHFFFAOYSA-N
23070
OC1=CC=CN=C1Br
C5H4BrNO
MFCD00006220
2-bromo-3-hydroxy pyridine, 2-bromo-3-hydroxy-pyridine, 2-bromo-3-hydroxypyridine, 2-bromo-3-hydroxypyridine 2-bromo-3-pyridinol, 2-bromo-3-pyridinol, 2-bromo-pyridin-3-ol, 2-bromo3-pyridinol, 3-hydroxy-2-bromopyridine, 3-pyridinol, 2-bromo, unii-9dab85h359
2-bromopyridin-3-ol

Specifications

6602-32-0
100.0
185°C to 188°C
C5H4BrNO
Br(C5H3N)OH
Glass bottle
2-bromo-3-hydroxy pyridine, 2-bromo-3-hydroxy-pyridine, 2-bromo-3-hydroxypyridine, 2-bromo-3-hydroxypyridine 2-bromo-3-pyridinol, 2-bromo-3-pyridinol, 2-bromo-pyridin-3-ol, 2-bromo3-pyridinol, 3-hydroxy-2-bromopyridine, 3-pyridinol, 2-bromo, unii-9dab85h359
OC1=CC=CN=C1Br
174.00
174
99%
98.5
Brown to Gray
99%
Authentic
MFCD00006220
25g
YKHQFTANTNMYPP-UHFFFAOYSA-N
2-bromopyridin-3-ol
23070
Crystalline Powder
2-Bromo-3-pyridinol, 99%
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Harmful in contact with skin.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 229-547-5

RTECSNumber : UU7705000

TSCA : TSCA

SDS
Product Certifications

RUO – Research Use Only