missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Bromo-3-pyridinecarboxaldehyde, 96%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 399670050

 View more versions of this product

Catalog No. AC399670050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

2-Bromo-3-pyridinecarboxaldehyde
95.0
100.0°C (3.0 mmHg)
Glass bottle
MFCD04966945
2-bromonicotinaldehyde, 2-bromo-3-formylpyridine, 2-bromo-3-pyridinecarboxaldehyde, 2-bromopyridine-3-carboxaldehyde, 3-pyridinecarboxaldehyde, 2-bromo, pubchem5057, acmc-1bypv, 2-bromo-3-formyl pyridine, bromo-3-pyridine carbaldehyde, bromo-3-pyridinecarboxaldehyde
C1=CC(=C(N=C1)Br)C=O
186
186
128071-75-0
100.0
95% min. (HPLC)
C6H4BrNO
5g
GNFWMEFWZWXLIN-UHFFFAOYSA-N
2-bromopyridine-3-carbaldehyde
4714951
96%

Chemical Identifiers

128071-75-0
186
GNFWMEFWZWXLIN-UHFFFAOYSA-N
4714951
C1=CC(=C(N=C1)Br)C=O
C6H4BrNO
MFCD04966945
2-bromonicotinaldehyde, 2-bromo-3-formylpyridine, 2-bromo-3-pyridinecarboxaldehyde, 2-bromopyridine-3-carboxaldehyde, 3-pyridinecarboxaldehyde, 2-bromo, pubchem5057, acmc-1bypv, 2-bromo-3-formyl pyridine, bromo-3-pyridine carbaldehyde, bromo-3-pyridinecarboxaldehyde
2-bromopyridine-3-carbaldehyde
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only