missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Bromo-3-nitropyridine, TCI America™

$58.06 - $198.61

Chemical Identifiers

CAS 19755-53-4
Molecular Formula C5H3BrN2O2
Molecular Weight (g/mol) 202.995
MDL Number MFCD00955613
InChI Key ZCCUFLITCDHRMG-UHFFFAOYSA-N
Synonym 2-bromo-3-nitro-pyridine, pyridine, 2-bromo-3-nitro, pyridine, 2-bromonitro, pubchem5955, acmc-1bt4y, 2-bromo-3-nitro pyridine, #, ksc490s2t, 2-bromo-3-nitropyridine
PubChem CID 555054
IUPAC Name 2-bromo-3-nitropyridine
SMILES C1=CC(=C(N=C1)Br)[N+](=O)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B41931G View Documents TCI America
B41931G
1g Each for $58.06
 
B41935G View Documents TCI America
B41935G
5g Each for $198.61
 
Specifications

Chemical Identifiers

19755-53-4
202.995
ZCCUFLITCDHRMG-UHFFFAOYSA-N
555054
C1=CC(=C(N=C1)Br)[N+](=O)[O-]
C5H3BrN2O2
MFCD00955613
2-bromo-3-nitro-pyridine, pyridine, 2-bromo-3-nitro, pyridine, 2-bromonitro, pubchem5955, acmc-1bt4y, 2-bromo-3-nitro pyridine, #, ksc490s2t, 2-bromo-3-nitropyridine
2-bromo-3-nitropyridine

Specifications

Yellow
1g
C5H3BrN2O2
2-bromo-3-nitro-pyridine, pyridine, 2-bromo-3-nitro, pyridine, 2-bromonitro, pubchem5955, acmc-1bt4y, 2-bromo-3-nitro pyridine, #, ksc490s2t, 2-bromo-3-nitropyridine
C1=CC(=C(N=C1)Br)[N+](=O)[O-]
202.995
203.00
≥98.0% (GC)
124°C
19755-53-4
MFCD00955613
ZCCUFLITCDHRMG-UHFFFAOYSA-N
2-bromo-3-nitropyridine
555054
Crystal-Powder at 20°C
2-Bromo-3-nitropyridine
Safety and Handling

Safety and Handling

TSCA : No