Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromo-3-nitropyridine, 98%

$68.26 - $263.69

Chemical Identifiers

Chemical Name or Material 2-Bromo-3-nitropyridine
CAS 19755-53-4
Molecular Formula C5H3BrN2O2
Formula Weight 203
MDL Number MFCD00955613
Beilstein 124504
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL2001903

  SDS
alfa aesar™
L2001903
1g Each for $68.26

AAL2001906

  SDS
alfa aesar™
L2001906
5g Each for $263.69
Description & Specifications

Specifications

Formula Weight 203
Chemical Name or Material 2-Bromo-3-nitropyridine
MDL Number MFCD00955613
Assay 98%
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
CAS 19755-53-4
Beilstein 124504
Molecular Formula C5H3BrN2O2
Melting Point 122° to 125°C
Quantity 1g
Health Hazard 2 GHS H Statement
H302-H312-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 1 Warning
Safety and Handling

Warning

GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H302-H312-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.