Learn More
2-Bromo-3-nitrobenzoic acid, tech. 90%, Thermo Scientific Chemicals
$123.22 - $123.22
Chemical Identifiers
CAS | 573-54-6 |
---|---|
Molecular Formula | C7H4BrNO4 |
Molecular Weight (g/mol) | 246.016 |
MDL Number | MFCD00074899 |
InChI Key | WTDJEGSXLFHZPY-UHFFFAOYSA-N |
Synonym | 2-bromo-3-nitrobenzoic acid, benzoic acid, 2-bromo-3-nitro, 2-bromo-3-nitrobenzoicacid, 2-bromo-3-nitro-benzoic acid, pubchem4714, acmc-1an8y, ksc273m5t, benzoicacid, 2-bromo-3-nitro, 2-bromo-3-nitrobenzoic acid, technical grade |
PubChem CID | 68452 |
SMILES | C1=CC(=C(C(=C1)[N+](=O)[O-])Br)C(=O)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH6079503
|
Thermo Scientific Chemicals
H6079503 |
1 g |
Each for $123.22
|
|
|||||
Description
2-Bromo-3-nitrobenzoic acid is used in preparation of 3-substituted 5-nitroisocoumarins1 and 2-bromo-3-nitrobenzaldehyde.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Bromo-3-nitrobenzoic acid is used in preparation of 3-substituted 5-nitroisocoumarins1 and 2-bromo-3-nitrobenzaldehyde.
Notes
Keep container tightly sealed. Store in refrigerated conditions. Incompatible with oxidizing agents and bases.
Chemical Identifiers
573-54-6 | |
246.016 | |
WTDJEGSXLFHZPY-UHFFFAOYSA-N | |
68452 |
C7H4BrNO4 | |
MFCD00074899 | |
2-bromo-3-nitrobenzoic acid, benzoic acid, 2-bromo-3-nitro, 2-bromo-3-nitrobenzoicacid, 2-bromo-3-nitro-benzoic acid, pubchem4714, acmc-1an8y, ksc273m5t, benzoicacid, 2-bromo-3-nitro, 2-bromo-3-nitrobenzoic acid, technical grade | |
C1=CC(=C(C(=C1)[N+](=O)[O-])Br)C(=O)O |
Specifications
573-54-6 | |
C7H4BrNO4 | |
1 g | |
WTDJEGSXLFHZPY-UHFFFAOYSA-N | |
246.016 | |
246.02 | |
Technical |
184°C to 186°C | |
MFCD00074899 | |
2-bromo-3-nitrobenzoic acid, benzoic acid, 2-bromo-3-nitro, 2-bromo-3-nitrobenzoicacid, 2-bromo-3-nitro-benzoic acid, pubchem4714, acmc-1an8y, ksc273m5t, benzoicacid, 2-bromo-3-nitro, 2-bromo-3-nitrobenzoic acid, technical grade | |
C1=CC(=C(C(=C1)[N+](=O)[O-])Br)C(=O)O | |
68452 | |
90% | |
2-Bromo-3-nitrobenzoic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 209-356-3
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only