Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromo-3'-nitroacetophenone, 97%

Chemical Identifiers

Chemical Name or Material 2-Bromo-3'-nitroacetophenone
CAS 2227-64-7
Molecular Formula C8H6BrNO3
Formula Weight 244.05
MDL Number MFCD00024512
UN Number UN1759
Beilstein 610594
Synonym alpha-Bromo-3-nitroacetophenone; 3-Nitrophenacyl bromide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1378109

  SDS
alfa aesar™
A1378109
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1378118

  SDS
alfa aesar™
A1378118
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 244.05
Melting Point 92° to 95°C
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Beilstein 610594
Synonym alpha-Bromo-3-nitroacetophenone; 3-Nitrophenacyl bromide
CAS 2227-64-7
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
MDL Number MFCD00024512
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, N.O.S.
Quantity 10g
Health Hazard 1 Danger
Assay 97%
Molecular Formula C8H6BrNO3
Chemical Name or Material 2-Bromo-3'-nitroacetophenone
UN Number UN1759

Applications
2-Bromo-3′-nitroacetophenone is used to prepare 2-bromo-3′-nitroacetophenone and 2-hydroxy-ethyl-1-[(3-nitro-phenyl)-2-oxoethyl]-piperidinium bromide. It can be used in agrochemical, pharmaceutical and dyestuff field etc.

Solubility
Soluble in methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, Strong bases.

Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.