missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromo-3-methylthiophene, 97%

Manufacturer:  Alfa Aesar™ L1139314

 View more versions of this product

Catalog No. AAL1139314



Specifications

Specifications

2-Bromo-3-methylthiophene
173°C to 176°C
14282-76-9
MFCD00059741
105719
YYJBWYBULYUKMR-UHFFFAOYSA-N
2-bromo-3-methylthiophene
84314
97%
68°C (154°F)
25g
C5H5BrS
UN2810
1-bromo-2-methyl thiophene
CC1=C(SC=C1)Br
177.059
177.07

Chemical Identifiers

14282-76-9
177.059
YYJBWYBULYUKMR-UHFFFAOYSA-N
84314
CC1=C(SC=C1)Br
C5H5BrS
MFCD00059741
1-bromo-2-methyl thiophene
2-bromo-3-methylthiophene
Safety and Handling

Safety and Handling

GHS H Statement
H331-H227-H302-H312-H315-H319-H335
Toxic if inhaled.
Combustible liquid.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.

Recommended Storage : 2° to 8°C

Documents