Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromo-3-methylpyridine, 98+%

$94.97 - $94.97

Chemical Identifiers

Chemical Name or Material 2-Bromo-3-methylpyridine
CAS 3430-17-9
Molecular Formula C6H6BrN
Formula Weight 172.03
MDL Number MFCD00239380
Beilstein 107896
Synonym 2-Bromo-3-picoline
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1952206

 
alfa aesar™
L1952206
5g Each for $94.97
Description & Specifications

Specifications

Formula Weight 172.03
Chemical Name or Material 2-Bromo-3-methylpyridine
Assay ≥98%
Molecular Formula C6H6BrN
CAS 3430-17-9
Synonym 2-Bromo-3-picoline
Health Hazard 3 GHS P Statement
P261-P301+P310-P305+P351+P338-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
MDL Number MFCD00239380
Beilstein 107896
Boiling Point 218° to 219°C
Quantity 5g
Flash Point >110°C (230°F)
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H311-H332-H315-H319
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Safety and Handling

Danger

GHS P Statement
P261-P301+P310-P305+P351+P338-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H311-H332-H315-H319
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.