missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Bromo-3-methylbenzoic acid, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC398860050

 View more versions of this product

Catalog No. AC398860050

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

2-Bromo-3-methylbenzoic acid
96.0
Authentic
134° to 138°C
Off-white
≥96.0%
C8H7BrO2
215.04
53663-39-1
100.0
Glass bottle
5g
Powder
97%
BrC6H3(CH3)COOH
MFCD00079721

Chemical Identifiers

53663-39-1
100.0
C8H7BrO2
215.04
2-Bromo-3-methylbenzoic acid
96.0
97%
BrC6H3(CH3)COOH
MFCD00079721
≥96.0%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.