Please login to your online account to display your discounted pricing

2-Bromo-3-methylbenzoic acid, 97%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC398860010

 View more versions of this product

Catalog No. AC398860010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Powder
Assay ≥96.0%
Formula Weight 215.04
Quantity 1g
Color Off-white
Melting Point 134° to 138°C
CAS Max % 100.0
CAS Min % 96.0
Linear Formula BrC6H3(CH3)COOH
Infrared Spectrum Authentic
Packaging Glass bottle
Molecular Formula C8H7BrO2
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS 53663-39-1
Chemical Name or Material 2-Bromo-3-methylbenzoic acid
Assay Percent Range 97%
MDL Number MFCD00079721
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

Specifications

Chemical Name or Material 2-Bromo-3-methylbenzoic acid
CAS 53663-39-1
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C8H7BrO2
Linear Formula BrC6H3(CH3)COOH
Formula Weight 215.04
MDL Number MFCD00079721
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

SDS & Safety Documents