Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromo-3-methyl-5-nitropyridine, 97%

$71.58 - $237.05

Chemical Identifiers

Chemical Name or Material 2-Bromo-3-methyl-5-nitropyridine
CAS 23132-21-0
Molecular Formula C6H5BrN2O2
Formula Weight 217.02
MDL Number MFCD03095065
Beilstein 1565441
Synonym 2-Bromo-5-nitro-3-picoline
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1985103

  SDS
alfa aesar™
L1985103
1g Each for $71.58

AAL1985106

  SDS
alfa aesar™
L1985106
5g Each for $237.05
Description & Specifications

Specifications

Formula Weight 217.02
Assay 97%
CAS 23132-21-0
Chemical Name or Material 2-Bromo-3-methyl-5-nitropyridine
Molecular Formula C6H5BrN2O2
MDL Number MFCD03095065
Melting Point 55° to 59°C
Beilstein 1565441
Synonym 2-Bromo-5-nitro-3-picoline
Quantity 1g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H312-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Safety and Handling

Warning

GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H302-H312-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.