missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Bromo-3'-methoxyacetophenone 97%, ACROS Organics™

Manufacturer:  Acros Organics 151910100

 View more versions of this product

Catalog No. AC151910100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

2-Bromo-3'-methoxyacetophenone
Glass bottle
59°C to 64°C
97%
96.0
97%
CH3OC6H4COCH2Br
2-bromo-1-3-methoxyphenyl ethanone, 2-bromo-3'-methoxyacetophenone, m-methoxyphenacyl bromide, 3'-methoxyphenacyl bromide, 3-bromoacetyl anisole, 2-bromo-1-3-methoxyphenyl ethan-1-one, 3-methoxyphenacyl bromide, bromomethyl 3-methoxyphenyl ketone, ccris 7998, 3-methoxyphenacylbromide
COC1=CC=CC(=C1)C(=O)CBr
229.073
229.07
≥96.0%
Authentic
Green-Yellow to White
10g
5000-65-7
100.0
C9H9BrO2
MFCD00000199
IOOHBIFQNQQUFI-UHFFFAOYSA-N
2-bromo-1-(3-methoxyphenyl)ethanone
101294
Crystalline Powder

Chemical Identifiers

5000-65-7
229.073
IOOHBIFQNQQUFI-UHFFFAOYSA-N
101294
COC1=CC=CC(=C1)C(=O)CBr
C9H9BrO2
MFCD00000199
2-bromo-1-3-methoxyphenyl ethanone, 2-bromo-3'-methoxyacetophenone, m-methoxyphenacyl bromide, 3'-methoxyphenacyl bromide, 3-bromoacetyl anisole, 2-bromo-1-3-methoxyphenyl ethan-1-one, 3-methoxyphenacyl bromide, bromomethyl 3-methoxyphenyl ketone, ccris 7998, 3-methoxyphenacylbromide
2-bromo-1-(3-methoxyphenyl)ethanone
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 225-666-1

TSCA : TSCA

SDS Documents