Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromo-3-iodo-5-methylpyridine, 95%

$172.39 - $172.39

Chemical Identifiers

Chemical Name or Material 2-Bromo-3-iodo-5-methylpyridine
CAS 65550-82-5
Molecular Formula C6H5BrIN
Formula Weight 297.92g/mol
MDL Number MFCD09037463
Synonym 6-Bromo-5-iodo-3-picoline
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2736203

 
alfa aesar™
H27362-03
1g Each for $172.39
Description & Specifications

Specifications

Melting Point 84° to 86°C
Quantity 1g
Chemical Name or Material 2-Bromo-3-iodo-5-methylpyridine
CAS 65550-82-5
Assay 95%
Molecular Formula C6H5BrIN
Formula Weight 297.92g/mol
MDL Number MFCD09037463
Synonym 6-Bromo-5-iodo-3-picoline
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.