Please login to your online account to display your discounted pricing

2-Bromo-3'-hydroxyacetophenone, 96%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC428620050

 View more versions of this product

Catalog No. AC428620050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 215.04
Quantity 5g
CAS Min % 95.0
CAS Max % 100.0
Chemical Name or Material 2-Bromo-3';-hydroxyacetophenone
Packaging Glass bottle
Assay Percent Range 96%
Molecular Formula C8H7BrO2
Linear Formula (HO)C6H4COCH2Br
MDL Number MFCD05664326
CAS 2491-37-4
Synonym 3-Hydroxyphenacyl bromide
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 2-Bromo-3';-hydroxyacetophenone
CAS 2491-37-4
CAS Min % 95.0
CAS Max % 100.0
Assay Percent Range 96%
Molecular Formula C8H7BrO2
Linear Formula (HO)C6H4COCH2Br
Formula Weight 215.04
MDL Number MFCD05664326
Synonym 3-Hydroxyphenacyl bromide
Accessories

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.