missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Alfa Aesar™ 2-Bromo-3-hexylthiophene, 98%
Manufacturer: Alfa Aesar™ H5624306
Description
Applications2-Bromo-3-hexylthiophene is used in the synthesis of End-capped Regioregular Poly(3-hexylthiophene). It is also used to synthesize well-defined head-to-tail-type oligothiophenes which are used in a number of high-technology applications including OLEDs.
Solubility
Slightly miscible with water.
Notes
Incompatible with strong oxidizing agentsstrong acids and bases.
Specifications
2-Bromo-3-hexylthiophene | |
110°C (230°F) | |
5g | |
C10H15BrS | |
2-bromanyl-3-hexyl-thiophene, 2-bromo-3-hexyl thiophene, 2-bromo-3-hexyl-thiophene, 2-bromo-3-hexylthiophene, 2-bromo-3-n-hexylthiophene, acmc-209o6r, amth047, ksc491i5d, thiophene, 2-bromo-3-hexyl | |
XQJNXCHDODCAJF-UHFFFAOYSA-N | |
2-bromo-3-hexylthiophene | |
10960141 | |
98% |
1.24 | |
1.529 | |
69249-61-2 | |
MFCD09907959 | |
Slightly miscible with water. | |
CCCCCCC1=C(SC=C1)Br | |
247.194 | |
247.2 |
Chemical Identifiers
69249-61-2 | |
247.194 | |
XQJNXCHDODCAJF-UHFFFAOYSA-N | |
10960141 | |
CCCCCCC1=C(SC=C1)Br |
C10H15BrS | |
MFCD09907959 | |
2-bromanyl-3-hexyl-thiophene, 2-bromo-3-hexyl thiophene, 2-bromo-3-hexyl-thiophene, 2-bromo-3-hexylthiophene, 2-bromo-3-n-hexylthiophene, acmc-209o6r, amth047, ksc491i5d, thiophene, 2-bromo-3-hexyl | |
2-bromo-3-hexylthiophene |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures