missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Bromo-3-furoic acid, 97%, Maybridge

$107.04 - $107.04

Chemical Identifiers

CAS 197846-05-2
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C5H3BrO3
Formula Weight 190.98
MDL Number MFCD09702417
Chemical Name or Material 2-Bromo-3-furoic acid
Assay 97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
CC64701DE SDS Fisher Scientific Company
CC64701DE
Amber Glass Bottle 5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC64701DA SDS Fisher Scientific Company
CC64701DA
Amber Glass Bottle 1g Each for $107.04
Specifications Description & Specifications

Chemical Identifiers

197846-05-2
100.0
C5H3BrO3
MFCD09702417
97%
96.5
97%
190.98
2-Bromo-3-furoic acid

Specifications

190.98
100.0
97%
MFCD09702417
Amber Glass Bottle
97%
5g
197846-05-2
C5H3BrO3
96.5
2-Bromo-3-furoic acid

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.