Learn More
2-Bromo-3-fluoropyridine, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H3198103
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2-Bromo-3-fluoropyridine | |
C5H3BrFN | |
MFCD04114128 | |
UN2810 | |
IFGLECYAEGYLSJ-UHFFFAOYSA-N | |
2-bromo-3-fluoropyridine | |
2762803 | |
97% |
40273-45-8 | |
1.545 | |
1 g | |
2-bromo-3-fluoro-pyridine, pyridine, 2-bromo-3-fluoro, abbypharma ap-17-5102, pubchem3002, 2-bromo-fluoropyridine, bromo-3-fluoro pyrdine, 2-bromo-3-fluoropyrdine, acmc-1am6d, ksc235k8b | |
C1=CC(=C(N=C1)Br)F | |
175.988 | |
175.99 |
Chemical Identifiers
40273-45-8 | |
175.988 | |
IFGLECYAEGYLSJ-UHFFFAOYSA-N | |
2762803 | |
C1=CC(=C(N=C1)Br)F |
C5H3BrFN | |
MFCD04114128 | |
2-bromo-3-fluoro-pyridine, pyridine, 2-bromo-3-fluoro, abbypharma ap-17-5102, pubchem3002, 2-bromo-fluoropyridine, bromo-3-fluoro pyrdine, 2-bromo-3-fluoropyrdine, acmc-1am6d, ksc235k8b | |
2-bromo-3-fluoropyridine |
Safety and Handling
GHS H Statement
H301-H311-H227-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Combustible liquid.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P235-P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P361-P363-P370+P378q-P501c
H227-H301+H311-H315-H319-H332-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only