Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromo-3-fluorobenzotrifluoride, 97%

$364.64 - $364.64

Chemical Identifiers

Chemical Name or Material 2-Bromo-3-fluorobenzotrifluoride
CAS 104540-42-3
Molecular Formula C7H3BrF4
Formula Weight 243
MDL Number MFCD00061243
Synonym 2-Bromo-1-fluoro-3-(trifluoromethyl)benzene; 2-Fluoro-6-(trifluoromethyl)bromobenzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2331206

  SDS
alfa aesar™
B2331206
5g Each for $364.64
Description & Specifications

Specifications

Formula Weight 243
Assay 97%
Chemical Name or Material 2-Bromo-3-fluorobenzotrifluoride
Health Hazard 3 GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 2 GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Molecular Formula C7H3BrF4
Synonym 2-Bromo-1-fluoro-3-(trifluoromethyl)benzene; 2-Fluoro-6-(trifluoromethyl)bromobenzene
Health Hazard 1 Warning
CAS 104540-42-3
Quantity 5g
MDL Number MFCD00061243
Safety and Handling

Warning

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.