missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Bromo-3-fluorobenzoic Acid 96.0+%, TCI America™
$126.08 - $391.52
Chemical Identifiers
CAS | 132715-69-6 |
---|---|
Molecular Formula | C7H4BrFO2 |
Molecular Weight (g/mol) | 219.009 |
MDL Number | MFCD01569398 |
InChI Key | KQRCBMPPEPNNDS-UHFFFAOYSA-N |
Synonym | 3-fluoro-2-bromobenzoic acid, 2-bromo-3-fluorobenzoicacid, 2-bromo-3-fluoro-benzoic acid, buttpark 22\01-97, benzoic acid, 2-bromo-3-fluoro, pubchem1319, 2-bromo-3-fluorobenzoic, acmc-1c0re, intermediates-zcf02016, ksc174c5p |
PubChem CID | 302622 |
IUPAC Name | 2-bromo-3-fluorobenzoic acid |
SMILES | C1=CC(=C(C(=C1)F)Br)C(=O)O |
Chemical Identifiers
132715-69-6 | |
219.009 | |
KQRCBMPPEPNNDS-UHFFFAOYSA-N | |
302622 | |
C1=CC(=C(C(=C1)F)Br)C(=O)O |
C7H4BrFO2 | |
MFCD01569398 | |
3-fluoro-2-bromobenzoic acid, 2-bromo-3-fluorobenzoicacid, 2-bromo-3-fluoro-benzoic acid, buttpark 22\01-97, benzoic acid, 2-bromo-3-fluoro, pubchem1319, 2-bromo-3-fluorobenzoic, acmc-1c0re, intermediates-zcf02016, ksc174c5p | |
2-bromo-3-fluorobenzoic acid |
Specifications
132715-69-6 | |
C7H4BrFO2 | |
1 g | |
KQRCBMPPEPNNDS-UHFFFAOYSA-N | |
2-bromo-3-fluorobenzoic acid | |
302622 | |
≥96.0% (GC,T) | |
2-Bromo-3-fluorobenzoic Acid |
160°C | |
MFCD01569398 | |
3-fluoro-2-bromobenzoic acid, 2-bromo-3-fluorobenzoicacid, 2-bromo-3-fluoro-benzoic acid, buttpark 22\01-97, benzoic acid, 2-bromo-3-fluoro, pubchem1319, 2-bromo-3-fluorobenzoic, acmc-1c0re, intermediates-zcf02016, ksc174c5p | |
C1=CC(=C(C(=C1)F)Br)C(=O)O | |
219.009 | |
219.01 | |
Crystalline Powder |
Safety and Handling
TSCA : No