missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromo-3-fluoroaniline, 97%

Manufacturer:  Alfa Aesar™ H61687MD

 View more versions of this product

Catalog No. AAH61687MD



Description

Description

Applications
2-Bromo-3-fluoroaniline is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Specifications

2-Bromo-3-fluoroaniline
94°C (201°F)
White
111721-75-6
MFCD07369915
2-bromo-3-fluoro-aniline
XZRSXRUYZXBTGD-UHFFFAOYSA-N
2-bromo-3-fluoroaniline
14770005
Crystalline
1.670 g/mL
1.583
250mg
C6H5BrFN
UN2811
Slightly soluble in water.
C1=CC(=C(C(=C1)F)Br)N
190.015
190.01
97%

Chemical Identifiers

111721-75-6
190.015
XZRSXRUYZXBTGD-UHFFFAOYSA-N
14770005
C1=CC(=C(C(=C1)F)Br)N
C6H5BrFN
MFCD07369915
2-bromo-3-fluoro-aniline
2-bromo-3-fluoroaniline
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

Documents