missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Bromo-3-cyclohexylthiophene, TCI America™

$319.31 - $959.47

Chemical Identifiers

CAS 241477-71-4
Molecular Formula C10H13BrS
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B54201G View Documents TCI America
B54201G
1g Each for $319.31
 
B54205G View Documents TCI America
B54205G
5g Each for $959.47
 
Specifications

Chemical Identifiers

241477-71-4
C10H13BrS

Specifications

Yellow
241477-71-4
245.18
≥95.0% (GC)
1g
C10H13BrS
Clear Liquid at 20°C
2-Bromo-3-cyclohexylthiophene
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator