missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Bromo-3-chloropyridine, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC442810010

 View more versions of this product

Catalog No. AC442810010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

2-Bromo-3-chloropyridine
60.0°C to 62.0°C
96424-68-9
C5H3BrClN
GOHBBINNYAWQGO-UHFFFAOYSA-N
2-bromo-3-chloropyridine
7016326
97%
Glass bottle
1g
96% min. (GC)
2-bromo-3-chloro-pyridine, 2-bromo-3-chloropyridine, 2-bromo-3-pyridyl chloride, 3-chloro-2-bromopyridine, abbypharma ap-18-5389, chloro bromo pyridine, ksc486m1b, pubchem5776, pyridine, 2-bromo-3-chloro
C1=CC(=C(N=C1)Br)Cl
192.44
192.44

Chemical Identifiers

96424-68-9
192.44
2-bromo-3-chloro-pyridine, 2-bromo-3-chloropyridine, 2-bromo-3-pyridyl chloride, 3-chloro-2-bromopyridine, abbypharma ap-18-5389, chloro bromo pyridine, ksc486m1b, pubchem5776, pyridine, 2-bromo-3-chloro
2-bromo-3-chloropyridine
C5H3BrClN
GOHBBINNYAWQGO-UHFFFAOYSA-N
7016326
C1=CC(=C(N=C1)Br)Cl
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and kee

GHS Signal Word: Warning

SDS Documents