missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromo-3-chlorobenzotrifluoride, 98%

$28.84 - $337.73

Chemical Identifiers

CAS 384-16-7
Molecular Formula C7H3BrClF3
Molecular Weight (g/mol) 259.45
MDL Number MFCD00673988
InChI Key GMUWNTUONIQRFP-UHFFFAOYSA-N
Synonym 2-bromo-1-chloro-3-trifluoromethyl benzene, 2-bromo-3-chlorobenzotrifluoride, 2-bromo-3-chloro-1-trifluoromethyl benzene, 2-bromanyl-1-chloranyl-3-trifluoromethyl benzene, benzene, 2-bromo-1-chloro-3-trifluoromethyl
PubChem CID 17750574
IUPAC Name 2-bromo-1-chloro-3-(trifluoromethyl)benzene
SMILES C1=CC(=C(C(=C1)Cl)Br)C(F)(F)F
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH64302MD View Documents Alfa Aesar™
H64302MD
250mg Each for $28.84
 
AAH6430203 View Documents Alfa Aesar™
H6430203
1g Each for $85.14
 
AAH6430206 View Documents Alfa Aesar™
H6430206
5g Each for $337.73
 
Specifications

Chemical Identifiers

384-16-7
259.45
GMUWNTUONIQRFP-UHFFFAOYSA-N
17750574
C1=CC(=C(C(=C1)Cl)Br)C(F)(F)F
C7H3BrClF3
MFCD00673988
2-bromo-1-chloro-3-trifluoromethyl benzene, 2-bromo-3-chlorobenzotrifluoride, 2-bromo-3-chloro-1-trifluoromethyl benzene, 2-bromanyl-1-chloranyl-3-trifluoromethyl benzene, benzene, 2-bromo-1-chloro-3-trifluoromethyl
2-bromo-1-chloro-3-(trifluoromethyl)benzene

Specifications

250mg
C7H3BrClF3
21501675
GMUWNTUONIQRFP-UHFFFAOYSA-N
2-bromo-1-chloro-3-(trifluoromethyl)benzene
17750574
Solid
2-Bromo-3-chlorobenzotrifluoride
384-16-7
MFCD00673988
2-bromo-1-chloro-3-trifluoromethyl benzene, 2-bromo-3-chlorobenzotrifluoride, 2-bromo-3-chloro-1-trifluoromethyl benzene, 2-bromanyl-1-chloranyl-3-trifluoromethyl benzene, benzene, 2-bromo-1-chloro-3-trifluoromethyl
C1=CC(=C(C(=C1)Cl)Br)C(F)(F)F
259.45
259.46
98%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning