Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromo-3'-chloroacetophenone, 97%

Chemical Identifiers

Chemical Name or Material 2-Bromo-3'-chloroacetophenone
CAS 41011-01-2
Formula Weight 233.49
MDL Number MFCD00792669
UN Number UN3335
Beilstein 2086002
Synonym 3-Chlorophenacyl bromide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6607703

  SDS
alfa aesar™
H6607703
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6607706

  SDS
alfa aesar™
H6607706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6607714

  SDS
alfa aesar™
H6607714
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Boiling Point 122° to 124°C
Melting Point 39° to 43°C
Formula Weight 233.49
MDL Number MFCD00792669
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Synonym 3-Chlorophenacyl bromide
Assay 97%
CAS 41011-01-2
Quantity 1g
Chemical Name or Material 2-Bromo-3'-chloroacetophenone
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
UN Number UN3335
DOT Information Transport Hazard Class: 9; Packaging Group: ; Proper Shipping Name: AVIATION REGULATED SOLID, N.O.S.
Beilstein 2086002
Storage Note 1 2° to 8°C
Health Hazard 1 Warning
Physical Form Solid

Applications
2-Bromo-3′-chloroacetophenone acts as a Substrate used in a one-step preparation of symmetrical 1,4-diketones in the presence of Zn-I2.

Notes
Store at 4°C. Incompatible with oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C