Learn More
2-Bromo-3'-chloroacetophenone, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H6607706
Description
2-Bromo-3'-chloroacetophenone acts as a Substrate used in a one-step preparation of symmetrical 1,4-diketones in the presence of Zn-I2.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Bromo-3′-chloroacetophenone acts as a Substrate used in a one-step preparation of symmetrical 1,4-diketones in the presence of Zn-I2.
Notes
Store at 4°C. Incompatible with oxidizing agents.
Specifications
2-Bromo-3'-chloroacetophenone | |
39°C to 43°C | |
>110°C (230°F) | |
MFCD00792669 | |
UN3335 | |
2-bromo-1-3-chlorophenyl ethanone, 3-chlorophenacyl bromide, 2-bromo-3'-chloroacetophenone, 2-bromo-1-3-chlorophenyl ethan-1-one, 2-bromo-1-3-chlorophenyl-1-ethanone, chlorophenacyl bromide, 3-chlorophenacylbromide, 3'-chlorophenacyl bromide, unii-n62718dtbs, ethanone, 2-bromo-1-3-chlorophenyl | |
C1=CC(=CC(=C1)Cl)C(=O)CBr | |
233.489 | |
233.49 | |
Solid |
41011-01-2 | |
122°C to 124°C | |
C8H6BrClO | |
5 g | |
2086002 | |
KJVRURZDIOVSSQ-UHFFFAOYSA-N | |
2-bromo-1-(3-chlorophenyl)ethanone | |
38738 | |
97% |
Chemical Identifiers
41011-01-2 | |
233.489 | |
KJVRURZDIOVSSQ-UHFFFAOYSA-N | |
38738 | |
C1=CC(=CC(=C1)Cl)C(=O)CBr |
C8H6BrClO | |
MFCD00792669 | |
2-bromo-1-3-chlorophenyl ethanone, 3-chlorophenacyl bromide, 2-bromo-3'-chloroacetophenone, 2-bromo-1-3-chlorophenyl ethan-1-one, 2-bromo-1-3-chlorophenyl-1-ethanone, chlorophenacyl bromide, 3-chlorophenacylbromide, 3'-chlorophenacyl bromide, unii-n62718dtbs, ethanone, 2-bromo-1-3-chlorophenyl | |
2-bromo-1-(3-chlorophenyl)ethanone |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
DOTInformation : Hazard Class: 9; Packaging Group: III
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only