Please login to your online account to display your discounted pricing

2-Bromo-3-chloro-5-(trifluoromethyl)pyridine, 97%, Maybridge


Quantity 10g
Packaging Amber glass bottle

Manufacturer: fisher scientific company  BTB09453EA

 View more versions of this product

Catalog No. BTB09453EA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 260.44
Quantity 10g
Assay 97%
CAS Max % 100.0
Molecular Formula C6H2BrClF3N
Assay Percent Range 97%
CAS 75806-84-7
MDL Number MFCD00153072
CAS Min % 96.5
Packaging Amber glass bottle
Health Hazard 1 Warning
Chemical Name or Material 2-Bromo-3-chloro-5-(trifluoromethyl)pyridine
Health Hazard 2 GHS P Statement Causes skin irritation. Causes serious eye irritation.
Health Hazard 3 GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. Take off contaminated clothing and wash before reuse. IF IN EYES: Rinse cautio

Specifications

Chemical Name or Material 2-Bromo-3-chloro-5-(trifluoromethyl)pyridine
CAS 75806-84-7
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C6H2BrClF3N
Formula Weight 260.44
MDL Number MFCD00153072
Accessories

Warning

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. Take off contaminated clothing and wash before reuse. IF IN EYES: Rinse cautio

GHS P Statement Causes skin irritation. Causes serious eye irritation.