missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromo-3'-chloro-4'-fluoroacetophenone, 96%

Manufacturer:  Alfa Aesar™ H6185206

 View more versions of this product

Catalog No. AAH6185206



Specifications

Specifications

2-Bromo-3'-chloro-4'-fluoroacetophenone
5g
C8H5BrClFO
2-bromo-1-3-chloro-4-fluorophenyl ethan-1-one
C1=CC(=C(C=C1C(=O)CBr)Cl)F
251.479
251.48
96%
33°C to 36°C
63529-30-6
MFCD05154991
JOCPGHGWUUBURW-UHFFFAOYSA-N
2-bromo-1-(3-chloro-4-fluorophenyl)ethanone
2782785
Solid

Chemical Identifiers

63529-30-6
251.479
JOCPGHGWUUBURW-UHFFFAOYSA-N
2782785
C1=CC(=C(C=C1C(=O)CBr)Cl)F
C8H5BrClFO
MFCD05154991
2-bromo-1-3-chloro-4-fluorophenyl ethan-1-one
2-bromo-1-(3-chloro-4-fluorophenyl)ethanone
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

Documents