Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromo-3,5-dinitrobenzoic acid, 98%

Chemical Identifiers

Chemical Name or Material 2-Bromo-3,5-dinitrobenzoic acid
CAS 116529-60-3
Formula Weight 291.01
MDL Number MFCD00458630
Beilstein 2146483
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6474503

 
alfa aesar™
H6474503
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6474506

 
alfa aesar™
H6474506
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6474514

 
alfa aesar™
H6474514
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point 213° to 216°C
Formula Weight 291.01
Chemical Name or Material 2-Bromo-3,5-dinitrobenzoic acid
Color Off-white
Physical Form Solid
Assay 98%
Quantity 1g
Sensitivity Light Sensitive
MDL Number MFCD00458630
CAS 116529-60-3
Beilstein 2146483