Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromo-3,5-diiodopyridine, 95%

$131.23 - $298.34

Chemical Identifiers

Chemical Name or Material 2-Bromo-3,5-diiodopyridine
CAS 885952-14-7
Molecular Formula C5H2BrI2N
Formula Weight 409.79
MDL Number MFCD06798228
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2786903

  SDS
alfa aesar™
H2786903
1g Each for $298.34

AAH27869MD

  SDS
alfa aesar™
H27869MD
250mg Each for $131.23
Description & Specifications

Specifications

Formula Weight 409.79
Molecular Formula C5H2BrI2N
Chemical Name or Material 2-Bromo-3,5-diiodopyridine
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 1 Danger
CAS 885952-14-7
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
Assay 95%
MDL Number MFCD06798228
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.