missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Bromo-3,5-dichloropyridine, TCI America™

$38.19 - $111.53

Chemical Identifiers

CAS 14482-51-0
Molecular Formula C5H2BrCl2N
Molecular Weight (g/mol) 226.882
MDL Number MFCD00233990
InChI Key QCKPJWDIDCGQRB-UHFFFAOYSA-N
Synonym 3,5-dichloro-2-bromopyridine, 2-bromo-3,5-dichloro-pyridine, pyridine, 2-bromo-3,5-dichloro, pubchem10223, acmc-1bqk7, 2-bromo-3,5-dichloropyridin, ksc494o1r, tpc-py058, 2-bromo-3,5 dichloro pyridine
PubChem CID 817090
IUPAC Name 2-bromo-3,5-dichloropyridine
SMILES C1=C(C=NC(=C1Cl)Br)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B37345G View Documents TCI America
B37345G
5g Each for $38.19
 
B373425G View Documents TCI America
B373425G
25g Each for $111.53
 
Specifications

Chemical Identifiers

14482-51-0
226.882
QCKPJWDIDCGQRB-UHFFFAOYSA-N
817090
C1=C(C=NC(=C1Cl)Br)Cl
C5H2BrCl2N
MFCD00233990
3,5-dichloro-2-bromopyridine, 2-bromo-3,5-dichloro-pyridine, pyridine, 2-bromo-3,5-dichloro, pubchem10223, acmc-1bqk7, 2-bromo-3,5-dichloropyridin, ksc494o1r, tpc-py058, 2-bromo-3,5 dichloro pyridine
2-bromo-3,5-dichloropyridine

Specifications

Undesignated
5g
C5H2BrCl2N
3,5-dichloro-2-bromopyridine, 2-bromo-3,5-dichloro-pyridine, pyridine, 2-bromo-3,5-dichloro, pubchem10223, acmc-1bqk7, 2-bromo-3,5-dichloropyridin, ksc494o1r, tpc-py058, 2-bromo-3,5 dichloro pyridine
C1=C(C=NC(=C1Cl)Br)Cl
226.882
226.88
≥98.0% (GC)
43°C
14482-51-0
MFCD00233990
QCKPJWDIDCGQRB-UHFFFAOYSA-N
2-bromo-3,5-dichloropyridine
817090
Crystal-Powder at 20°C
2-Bromo-3,5-dichloropyridine
Safety and Handling

Safety and Handling

EINECSNumber : (5)-5257

TSCA : No