missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Bromo-3,5-dichloropyridine 98.0+%, TCI America™

Supplier:  TCI America B373425G

 View more versions of this product

Catalog No. B373425G



Description

Description

Specifications

Specifications

2-Bromo-3,5-dichloropyridine
43°C
MFCD00233990
3,5-dichloro-2-bromopyridine, 2-bromo-3,5-dichloro-pyridine, pyridine, 2-bromo-3,5-dichloro, pubchem10223, acmc-1bqk7, 2-bromo-3,5-dichloropyridin, ksc494o1r, tpc-py058, 2-bromo-3,5 dichloro pyridine
ClC1=CN=C(Br)C(Cl)=C1
226.88
226.88
Crystalline Powder
14482-51-0
C5H2BrCl2N
25 g
QCKPJWDIDCGQRB-UHFFFAOYSA-N
2-bromo-3,5-dichloropyridine
817090
≥98.0% (GC)

Chemical Identifiers

14482-51-0
226.88
QCKPJWDIDCGQRB-UHFFFAOYSA-N
817090
ClC1=CN=C(Br)C(Cl)=C1
C5H2BrCl2N
MFCD00233990
3,5-dichloro-2-bromopyridine, 2-bromo-3,5-dichloro-pyridine, pyridine, 2-bromo-3,5-dichloro, pubchem10223, acmc-1bqk7, 2-bromo-3,5-dichloropyridin, ksc494o1r, tpc-py058, 2-bromo-3,5 dichloro pyridine
2-bromo-3,5-dichloropyridine
Safety and Handling

Safety and Handling

EINECSNumber : (5)-5257

TSCA : No

SDS
Documents

Documents

Promotions

Promotions