missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromo-3,5-dichloropyridine, 98%

Manufacturer:  Alfa Aesar™ H6113822

 View more versions of this product

Catalog No. AAH6113822



Specifications

Specifications

2-Bromo-3,5-dichloropyridine
100g
C5H2BrCl2N
UN2811
QCKPJWDIDCGQRB-UHFFFAOYSA-N
2-bromo-3,5-dichloropyridine
817090
Solid
Brown
14482-51-0
MFCD00233990
2-bromo-3,5 dichloro pyridine
C1=C(C=NC(=C1Cl)Br)Cl
226.882
226.89
98%

Chemical Identifiers

14482-51-0
226.882
QCKPJWDIDCGQRB-UHFFFAOYSA-N
817090
C1=C(C=NC(=C1Cl)Br)Cl
C5H2BrCl2N
MFCD00233990
2-bromo-3,5 dichloro pyridine
2-bromo-3,5-dichloropyridine
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

Documents