missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromo-3,5-dichloropyridine-4-carboxylic acid, 95%

$35.93 - $422.54

Chemical Identifiers

CAS 343781-56-6
Molecular Formula C6H2BrCl2NO2
Molecular Weight (g/mol) 270.891
MDL Number MFCD03411637
InChI Key DSKHVZJZZXHWPU-UHFFFAOYSA-N
Synonym 2-bromo-3,5-dichloroisonicotinic acid, pubchem13557, 2-bromo-3,5-dichloro-isonicotinic acid, 4-pyridinecarboxylicacid, 2-bromo-3,5-dichloro, 2-bromo-3,5-dichloroisonicotinic acid
PubChem CID 2762469
IUPAC Name 2-bromo-3,5-dichloropyridine-4-carboxylic acid
SMILES C1=C(C(=C(C(=N1)Br)Cl)C(=O)O)Cl
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH63739MD View Documents Alfa Aesar™
H63739MD
250mg Each for $35.93
 
AAH6373903 View Documents Alfa Aesar™
H6373903
1g Each for $106.26
 
AAH6373906 View Documents Alfa Aesar™
H6373906
5g Each for $422.54
 
Specifications

Chemical Identifiers

343781-56-6
270.891
DSKHVZJZZXHWPU-UHFFFAOYSA-N
2762469
C1=C(C(=C(C(=N1)Br)Cl)C(=O)O)Cl
C6H2BrCl2NO2
MFCD03411637
2-bromo-3,5-dichloroisonicotinic acid, pubchem13557, 2-bromo-3,5-dichloro-isonicotinic acid, 4-pyridinecarboxylicacid, 2-bromo-3,5-dichloro, 2-bromo-3,5-dichloroisonicotinic acid
2-bromo-3,5-dichloropyridine-4-carboxylic acid

Specifications

White
343781-56-6
MFCD03411637
DSKHVZJZZXHWPU-UHFFFAOYSA-N
2-bromo-3,5-dichloropyridine-4-carboxylic acid
2762469
Solid
2-Bromo-3,5-dichloropyridine-4-carboxylic acid
250mg
C6H2BrCl2NO2
2-bromo-3,5-dichloroisonicotinic acid, pubchem13557, 2-bromo-3,5-dichloro-isonicotinic acid, 4-pyridinecarboxylicacid, 2-bromo-3,5-dichloro, 2-bromo-3,5-dichloroisonicotinic acid
C1=C(C(=C(C(=N1)Br)Cl)C(=O)O)Cl
270.891
270.9
95%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning