Alfa Aesar™ 2-Bromo-3,5-dichloropyridine-4-carboxylic acid, 95%
Manufacturer:  Alfa Aesar™ H63739MD
 View more versions of this product
Catalog No. AAH63739MD
Specifications
Specifications
2-Bromo-3,5-dichloropyridine-4-carboxylic acid
250mg
C6H2BrCl2NO2
2-bromo-3,5-dichloroisonicotinic acid, pubchem13557, 2-bromo-3,5-dichloro-isonicotinic acid, 4-pyridinecarboxylicacid, 2-bromo-3,5-dichloro, 2-bromo-3,5-dichloroisonicotinic acid
C1=C(C(=C(C(=N1)Br)Cl)C(=O)O)Cl
270.891
270.9
95%
White
343781-56-6
MFCD03411637
DSKHVZJZZXHWPU-UHFFFAOYSA-N
2-bromo-3,5-dichloropyridine-4-carboxylic acid
2762469
Solid
Chemical Identifiers
343781-56-6
270.891
DSKHVZJZZXHWPU-UHFFFAOYSA-N
2762469
C1=C(C(=C(C(=N1)Br)Cl)C(=O)O)Cl
C6H2BrCl2NO2
MFCD03411637
2-bromo-3,5-dichloroisonicotinic acid, pubchem13557, 2-bromo-3,5-dichloro-isonicotinic acid, 4-pyridinecarboxylicacid, 2-bromo-3,5-dichloro, 2-bromo-3,5-dichloroisonicotinic acid
2-bromo-3,5-dichloropyridine-4-carboxylic acid
Safety and Handling
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
SDS Documents