Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromo-3',5'-di-tert-butyl-4'-hydroxyacetophenone, 97%

Chemical Identifiers

Chemical Name or Material 2-Bromo-3',5'-di-tert-butyl-4'-hydroxyacetophenone
CAS 14386-64-2
Molecular Formula C16H23BrO2
Formula Weight 327.26
MDL Number MFCD00173889
UN Number UN1759
Synonym 3,5-Di-tert-butyl-4-hydroxyphenacyl bromide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2683503

 
alfa aesar™
H2683503
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH2683506

 
alfa aesar™
H2683506
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH2683514

 
alfa aesar™
H2683514
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 327.26
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Chemical Name or Material 2-Bromo-3',5'-di-tert-butyl-4'-hydroxyacetophenone
Assay 97%
Molecular Formula C16H23BrO2
MDL Number MFCD00173889
CAS 14386-64-2
UN Number UN1759
Melting Point 104° to 107°C
Synonym 3,5-Di-tert-butyl-4-hydroxyphenacyl bromide
Quantity 1g
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, N.O.S.
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.