missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromo-3,4-difluoroaniline, 97%

Manufacturer:  Alfa Aesar™ H6187603

 View more versions of this product

Catalog No. AAH6187603



Specifications

Specifications

2-Bromo-3,4-difluoroaniline
1092349-87-5
MFCD11035924
2-bromo-3,4-difluoro aniline, 2-bromo-3,4-difluorophenylamine, 2-bromo-3,4-difluorobenzenamine
C1=CC(=C(C(=C1N)Br)F)F
208.006
208
97%
1g
C6H4BrF2N
UN2811
ZGZURPPCPVPDMX-UHFFFAOYSA-N
2-bromo-3,4-difluoroaniline
22731604
Solid

Chemical Identifiers

1092349-87-5
208.006
ZGZURPPCPVPDMX-UHFFFAOYSA-N
22731604
C1=CC(=C(C(=C1N)Br)F)F
C6H4BrF2N
MFCD11035924
2-bromo-3,4-difluoro aniline, 2-bromo-3,4-difluorophenylamine, 2-bromo-3,4-difluorobenzenamine
2-bromo-3,4-difluoroaniline
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

Documents