Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromo-3,4,5,6-tetrafluorobenzoyl chloride, 97%

Chemical Identifiers

Chemical Name or Material 2-Bromo-3,4,5,6-tetrafluorobenzoyl chloride
CAS 151096-42-3
Molecular Formula C7BrClF4O
Formula Weight 291.43
MDL Number MFCD03001174
UN Number UN3265
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL2015103

  SDS
alfa aesar™
L2015103
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL2015106

  SDS
alfa aesar™
L2015106
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL2015114

  SDS
alfa aesar™
L2015114
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 291.43
UN Number UN3265
Boiling Point 125° to 126°C (80mmHg)
Molecular Formula C7BrClF4O
CAS 151096-42-3
Assay 97%
MDL Number MFCD03001174
Quantity 1g
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
Chemical Name or Material 2-Bromo-3,4,5,6-tetrafluorobenzoyl chloride
Safety and Handling

Danger

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.