missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Bromo-3,3,3-trifluoro-1-propene, 97%, ACROS Organics™

Manufacturer:  Acros Organics AC434460050

 View more versions of this product

Catalog No. AC434460050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

2-Bromo-3, 3, 3-trifluoro-1-propene
Authentic
1.3520 to 1.3550 (20°C, 589nm)
Colorless
1514-82-5
100.0
C3H2BrF3
QKBKGNDTLQFSEU-UHFFFAOYSA-N
2-bromo-3,3,3-trifluoroprop-1-ene
272696
Liquid
−23°C
Glass bottle
29.0°C to 30.0°C
5g
96.0
97%
2-bromo-3,3,3-trifluoropropene, 2-bromo-3,3,3-trifluoro-1-propene, 1-propene, 2-bromo-3,3,3-trifluoro, 2-bromo-3,3,3-trifluoro-propene, 2-bromo-3,3,3-trifluoro-prop-1-ene, 3,3,3-trifluoro-2-bromo-1-propene, 2-bromotrifluoropropene, acmc-1bt5r, ksc495o1d, 3,3,3-trifluoro-2-bromopropene
C=C(C(F)(F)F)Br
174.948
174.95
97%

Chemical Identifiers

1514-82-5
174.948
2-bromo-3,3,3-trifluoropropene, 2-bromo-3,3,3-trifluoro-1-propene, 1-propene, 2-bromo-3,3,3-trifluoro, 2-bromo-3,3,3-trifluoro-propene, 2-bromo-3,3,3-trifluoro-prop-1-ene, 3,3,3-trifluoro-2-bromo-1-propene, 2-bromotrifluoropropene, acmc-1bt5r, ksc495o1d, 3,3,3-trifluoro-2-bromopropene
2-bromo-3,3,3-trifluoroprop-1-ene
C3H2BrF3
QKBKGNDTLQFSEU-UHFFFAOYSA-N
272696
C=C(C(F)(F)F)Br
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Harmful if inhaled.
Causes serious eye irritation.
Causes skin irritation.
Harmful in contact with skin.
Harmful if swallowed.
Extremely flammable liquid and vapor.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Gently wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS Documents