Learn More
2-Bromo-3,3,3-trifluoro-1-propene, 97%, Thermo Scientific Chemicals
$135.66 - $421.04
Chemical Identifiers
CAS | 1514-82-5 |
---|---|
Molecular Formula | C3H2BrF3 |
Molecular Weight (g/mol) | 174.95 |
MDL Number | MFCD00077469 |
InChI Key | QKBKGNDTLQFSEU-UHFFFAOYSA-N |
Synonym | 2-bromo-3,3,3-trifluoropropene, 2-bromo-3,3,3-trifluoro-1-propene, 1-propene, 2-bromo-3,3,3-trifluoro, 2-bromo-3,3,3-trifluoro-propene, 2-bromo-3,3,3-trifluoro-prop-1-ene, 3,3,3-trifluoro-2-bromo-1-propene, 2-bromotrifluoropropene, acmc-1bt5r, ksc495o1d, 3,3,3-trifluoro-2-bromopropene |
PubChem CID | 272696 |
IUPAC Name | 2-bromo-3,3,3-trifluoroprop-1-ene |
SMILES | FC(F)(F)C(Br)=C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAB2461406
|
Thermo Scientific Chemicals
B2461406 |
5 g |
Each for $135.66
|
|
|||||
AAB2461414
|
Thermo Scientific Chemicals
B2461414 |
25 g |
Each for $421.04
|
|
|||||
Description
It is used as intermediate in production of organ fluorine compounds & as a monomer in fluoropolymer production. It is used as a trifluoromethylated synthetic building block for the synthesis of trifluoromethyl-substituted molecules.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used as intermediate in production of organ fluorine compounds & as a monomer in fluoropolymer production. It is used as a trifluoromethylated synthetic building block for the synthesis of trifluoromethyl-substituted molecules.
Solubility
Soluble in most organic solvents, immiscible with water.
Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.
Chemical Identifiers
1514-82-5 | |
174.95 | |
QKBKGNDTLQFSEU-UHFFFAOYSA-N | |
272696 | |
FC(F)(F)C(Br)=C |
C3H2BrF3 | |
MFCD00077469 | |
2-bromo-3,3,3-trifluoropropene, 2-bromo-3,3,3-trifluoro-1-propene, 1-propene, 2-bromo-3,3,3-trifluoro, 2-bromo-3,3,3-trifluoro-propene, 2-bromo-3,3,3-trifluoro-prop-1-ene, 3,3,3-trifluoro-2-bromo-1-propene, 2-bromotrifluoropropene, acmc-1bt5r, ksc495o1d, 3,3,3-trifluoro-2-bromopropene | |
2-bromo-3,3,3-trifluoroprop-1-ene |
Specifications
1514-82-5 | |
1.353 | |
5 g | |
Soluble in most organic solvents,immiscible with water. | |
FC(F)(F)C(Br)=C | |
174.95 | |
174.95 | |
2-Bromo-3,3,3-trifluoro-1-propene |
C3H2BrF3 | |
MFCD00077469 | |
2-bromo-3,3,3-trifluoropropene, 2-bromo-3,3,3-trifluoro-1-propene, 1-propene, 2-bromo-3,3,3-trifluoro, 2-bromo-3,3,3-trifluoro-propene, 2-bromo-3,3,3-trifluoro-prop-1-ene, 3,3,3-trifluoro-2-bromo-1-propene, 2-bromotrifluoropropene, acmc-1bt5r, ksc495o1d, 3,3,3-trifluoro-2-bromopropene | |
QKBKGNDTLQFSEU-UHFFFAOYSA-N | |
2-bromo-3,3,3-trifluoroprop-1-ene | |
272696 | |
97% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P271-P304+P340-P312-P501c
H335-H336
TSCA : Yes
RUO – Research Use Only