Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromo-3-(2-tetrahydropyranyloxy)pyridine, 97%

$108.51 - $108.51

Chemical Identifiers

Chemical Name or Material 2-Bromo-3- (2-tetrahydropyranyloxy)pyridine
CAS 93560-60-2
Molecular Formula C10H12BrNO2
Formula Weight 258.12
MDL Number MFCD06411700
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4477403

  SDS
alfa aesar™
4477403
1g Each for $108.51
Description & Specifications

Specifications

Formula Weight 258.12
Chemical Name or Material 2-Bromo-3- (2-tetrahydropyranyloxy)pyridine
Physical Form Solid
Assay 97%
Molecular Formula C10H12BrNO2
MDL Number MFCD06411700
CAS 93560-60-2
Melting Point 67°C
Quantity 1g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.