Learn More
2-Bromo-2-nitropropane-1,3-diol, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1163922
Description
2-Bromo-2-nitropropane-1,3-diol Cosmetics, Preservatives for milk samples, Topical medications and also provide wide spectrum of antibacterial activity.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Bromo-2-nitropropane-1,3-diol Cosmetics, Preservatives for milk samples, Topical medications and also provide wide spectrum of antibacterial activity.
Solubility
Soluble in water(280g/L).
Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
2-Bromo-2-nitropropane-1,3-diol | |
∼122°C to 132°C | |
C3H6BrNO4 | |
100 g | |
1705868 | |
bronopol, 2-bromo-2-nitro-1,3-propanediol, bronosol, bronocot, bronidiol, bronopolu, bronotak, 1,3-propanediol, 2-bromo-2-nitro, onyxide 500, lexgard bronopol | |
LVDKZNITIUWNER-UHFFFAOYSA-N | |
2-bromo-2-nitropropane-1,3-diol | |
2450 | |
199.99 |
52-51-7 | |
5 to 7 | |
MFCD00007390 | |
UN3241 | |
14,1447 | |
Soluble in water(280g/L). | |
C(C(CO)([N+](=O)[O-])Br)O | |
199.988 | |
CHEBI:31306 | |
≥98% |
Chemical Identifiers
52-51-7 | |
199.988 | |
LVDKZNITIUWNER-UHFFFAOYSA-N | |
2450 | |
2-bromo-2-nitropropane-1,3-diol |
C3H6BrNO4 | |
MFCD00007390 | |
bronopol, 2-bromo-2-nitro-1,3-propanediol, bronosol, bronocot, bronidiol, bronopolu, bronotak, 1,3-propanediol, 2-bromo-2-nitro, onyxide 500, lexgard bronopol | |
CHEBI:31306 | |
C(C(CO)([N+](=O)[O-])Br)O |
Safety and Handling
GHS H Statement
H301-H331-H318-H228-H312-H315-H335
Toxic if swallowed.
Toxic if inhaled.
Causes serious eye damage.
Flammable solid.
Harmful in contact with skin.
Causes skin irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c
H302+H312-H315-H318-H335-H500
DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: 2-BROMO-2-NITROPROPANE-1,3-DIOL
EINECSNumber : 200-143-0
RTECSNumber : TY3385000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only