missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Bromo-2-nitro-1,3-propanediol, 98%, ACROS Organics™

$48.80 - $580.92

Chemical Identifiers

CAS 52-51-7
Molecular Formula C3H6BrNO4
Molecular Weight (g/mol) 199.988
MDL Number MFCD00007390
InChI Key LVDKZNITIUWNER-UHFFFAOYSA-N
Synonym bronopol, 2-bromo-2-nitro-1,3-propanediol, bronosol, bronocot, bronidiol, bronopolu, bronotak, 1,3-propanediol, 2-bromo-2-nitro, onyxide 500, lexgard bronopol
PubChem CID 2450
ChEBI CHEBI:31306
IUPAC Name 2-bromo-2-nitropropane-1,3-diol
SMILES C(C(CO)([N+](=O)[O-])Br)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC158825000 SDS View Documents Acros Organics
AC158825000
500g Plastic bottle Each for $580.92
 
AC158820250 SDS View Documents Acros Organics
158820250
25g Plastic bottle Each for $48.80
 
AC158821000 SDS View Documents Acros Organics
158821000
100g Plastic bottle Each for $136.83
 
Specifications

Chemical Identifiers

52-51-7
199.988
LVDKZNITIUWNER-UHFFFAOYSA-N
2450
2-bromo-2-nitropropane-1,3-diol
C3H6BrNO4
MFCD00007390
bronopol, 2-bromo-2-nitro-1,3-propanediol, bronosol, bronocot, bronidiol, bronopolu, bronotak, 1,3-propanediol, 2-bromo-2-nitro, onyxide 500, lexgard bronopol
CHEBI:31306
C(C(CO)([N+](=O)[O-])Br)O

Specifications

Authentic
0.5% max.
>120.0°C
52-51-7
100.0
C3H6BrNO4
MFCD00007390
15, 1457
LVDKZNITIUWNER-UHFFFAOYSA-N
2-bromo-2-nitropropane-1,3-diol
2450
199.99
98%
Plastic bottle
White to Yellow
500g
97.5
98%
HOCH2C(Br)(NO2)CH2OH
01, IV, 2501
bronopol, 2-bromo-2-nitro-1,3-propanediol, bronosol, bronocot, bronidiol, bronopolu, bronotak, 1,3-propanediol, 2-bromo-2-nitro, onyxide 500, lexgard bronopol
C(C(CO)([N+](=O)[O-])Br)O
199.988
CHEBI:31306
Crystals or Crystalline Powder
2-Bromo-2-nitro-1, 3-propanediol
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Harmful in contact with skin.
Causes serious eye damage.
Harmful if swallowed.
Causes skin irritation.
Very toxic to aquatic life.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger